Assay Details
| Gene Symbol: Klp3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297901.1 | NP_001284830.1 | 2 - 3 | 900 | 60 | |
| NM_080314.3 | NP_525053.1 | 2 - 3 | 1049 | 60 | ||
| GenBank mRNA | AF132186.1 | - | 2 - 3 | 832 | 60 | |
More Information
Set Membership: |
|

Klp3A
Genomic Map