Assay Details
| Gene Symbol: Klp3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297901.1 | NP_001284830.1 | 4 - 5 | 3320 | 61 | |
| NM_080314.3 | NP_525053.1 | 4 - 5 | 3469 | 61 | ||
| GenBank mRNA | AF132186.1 | - | 4 - 5 | 3252 | 61 | |
More Information
Set Membership: |
|

Klp3A
Genomic Map