Assay Details
| Gene Symbol: ovo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038742.3 | NP_001033831.1 | 2 - 3 | 3407 | 79 | |
| NM_080338.5 | NP_525077.2 | 3 - 4 | 3248 | 79 | ||
| GenBank mRNA | AY119649.1 | - | 3 - 4 | 3248 | 79 | |
More Information
Set Membership: |
|

ovo
Genomic Map