Assay Details
| Gene Symbol: Rok | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298406.1 | NP_001285335.1 | 9 - 10 | 4901 | 62 | |
| NM_001298407.1 | NP_001285336.1 | 9 - 10 | 4928 | 62 | ||
| NM_001298408.1 | NP_001285337.1 | 8 - 9 | 3929 | 62 | ||
| NM_080535.3 | NP_536796.2 | 9 - 10 | 4928 | 62 | ||
| GenBank mRNA | BT044181.1 | - | 9 - 10 | 4933 | 62 | |
More Information
Set Membership: |
|

Rok
Genomic Map