Assay Details
| Gene Symbol: CG8188 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038763.2 | NP_001033852.1 | 2 - 3 | 1045 | 54 | |
| NM_001298454.1 | NP_001285383.1 | 2 - 3 | 1045 | 54 | ||
| NM_001298455.1 | NP_001285384.1 | 2 - 3 | 1045 | 54 | ||
| NM_133009.3 | NP_573237.2 | 2 - 3 | 1292 | 54 | ||
| GenBank mRNA | BT031269.1 | - | 2 - 3 | 969 | 54 | |
| DQ151896.1 | - | 2 - 3 | 852 | 54 | ||
| DQ151897.1 | - | 2 - 3 | 958 | 54 | ||
More Information
Set Membership: |
|

CG8188
Genomic Map