Assay Details
| Gene Symbol: E2f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299186.1 | NP_001286115.1 | 1 - 2 | 1009 | 60 | |
| NM_058007.5 | NP_477355.1 | 1 - 2 | 492 | 60 | ||
| GenBank mRNA | AB016824.1 | - | 1 - 2 | 463 | 60 | |
| AY069118.1 | - | 1 - 2 | 491 | 60 | ||
More Information
Set Membership: |
|

E2f2
Genomic Map