Assay Details
| Gene Symbol: E2f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299186.1 | NP_001286115.1 | 3 - 4 | 1373 | 143 | |
| NM_058007.5 | NP_477355.1 | 3 - 4 | 856 | 143 | ||
| GenBank mRNA | AB016824.1 | - | 3 - 4 | 827 | 143 | |
| AY069118.1 | - | 3 - 4 | 855 | 143 | ||
More Information
Set Membership: |
|

E2f2
Genomic Map