Assay Details
| Gene Symbol: eIF3e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058037.5 | NP_477385.1 | 2 - 3 | 586 | 60 | |
| GenBank mRNA | AF132551.1 | - | 2 - 3 | 615 | 60 | |
| U89162.1 | - | 2 - 3 | 552 | 60 | ||
More Information
Set Membership: |
|

eIF3e
Genomic Map