Assay Details
| Gene Symbol: Sam-S | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_164361.2 | NP_722599.1 | 5 - 6 | 614 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sam-S| Gene Symbol: Sam-S | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_164361.2 | NP_722599.1 | 5 - 6 | 614 | 72 | |
Set Membership: |
|