Assay Details
| Gene Symbol: Gyc32E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103678.2 | NP_001097148.1 | 4 - 5 | 717 | 72 | |
| NM_001298905.1 | NP_001285834.1 | 4 - 5 | 717 | 72 | ||
| NM_176013.2 | NP_788027.1 | 4 - 5 | 717 | 72 | ||
More Information
Set Membership: |
|

Gyc32E
Genomic Map