Assay Details
| Gene Symbol: MED9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176220.2 | NP_788400.1 | 1 - 2 | 268 | 71 | |
| GenBank mRNA | KX531396.1 | - | 1 - 2 | 206 | 71 | |
| Gene Symbol: CG42518 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169716.2 | NP_001163187.1 | 1 - 2 | 268 | 71 | |
| NM_001169717.1 | NP_001163188.1 | 2 - 3 | 222 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001169716.2, NM_001169717.1 transcripts.
Set Membership: |
|

MED9
Genomic Map