Assay Details
| Gene Symbol: CG33143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299803.1 | NP_001286732.1 | 2 - 3 | 1750 | 84 | |
| NM_001299804.1 | NP_001286733.1 | 2 - 3 | 1750 | 84 | ||
| NM_001382063.1 | NP_001368934.1 | 2 - 3 | 1750 | 84 | ||
| NM_176246.2 | NP_788426.1 | 2 - 3 | 1750 | 84 | ||
| GenBank mRNA | AY095532.1 | - | 2 - 3 | 1748 | 84 | |
More Information
Set Membership: |
|

CG33143
Genomic Map