Assay Details
| Gene Symbol: CG33143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176246.2 | NP_788426.1 | 6 - 7 | 2398 | 57 | |
| GenBank mRNA | AY095532.1 | - | 6 - 7 | 2396 | 57 | |
More Information
Set Membership: |
|

CG33143
Genomic Map