Assay Details
| Gene Symbol: Mp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169885.2 | NP_001163356.1 | 15 - 16 | 2405 | 72 | |
| NM_001169886.2 | NP_001163357.1 | 10 - 11 | 2639 | 72 | ||
| NM_001169887.2 | NP_001163358.1 | 16 - 17 | 2440 | 72 | ||
| NM_001169888.2 | NP_001163359.1 | 12 - 13 | 2744 | 72 | ||
| NM_001169890.2 | NP_001163361.1 | 10 - 11 | 1602 | 72 | ||
| NM_001169891.2 | NP_001163362.2 | 15 - 16 | 2385 | 72 | ||
| NM_001169892.2 | NP_001163363.1 | 14 - 15 | 2299 | 72 | ||
| NM_001169893.2 | NP_001163364.1 | 15 - 16 | 2410 | 72 | ||
| NM_001300032.1 | NP_001286961.1 | 15 - 16 | 2410 | 72 | ||
| NM_001300033.1 | NP_001286962.1 | 13 - 14 | 2305 | 72 | ||
| NM_176293.3 | NP_788471.1 | 10 - 11 | 2639 | 72 | ||
| NM_176294.5 | NP_788472.2 | 10 - 11 | 2639 | 72 | ||
| GenBank mRNA | AY118310.1 | - | 10 - 11 | 2614 | 72 | |
More Information
Set Membership: |
|

Mp
Genomic Map