Assay Details
| Gene Symbol: Mp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169885.2 | NP_001163356.1 | 17 - 18 | 2652 | 63 | |
| NM_001169886.2 | NP_001163357.1 | 12 - 13 | 2886 | 63 | ||
| NM_001169887.2 | NP_001163358.1 | 18 - 19 | 2687 | 63 | ||
| NM_001169888.2 | NP_001163359.1 | 14 - 15 | 2991 | 63 | ||
| NM_001169890.2 | NP_001163361.1 | 12 - 13 | 1849 | 63 | ||
| NM_001169891.2 | NP_001163362.2 | 17 - 18 | 2632 | 63 | ||
| NM_001169892.2 | NP_001163363.1 | 16 - 17 | 2546 | 63 | ||
| NM_001169893.2 | NP_001163364.1 | 17 - 18 | 2657 | 63 | ||
| NM_001259722.1 | NP_001246651.1 | 18 - 19 | 2705 | 63 | ||
| NM_001300031.1 | NP_001286960.1 | 13 - 14 | 2964 | 63 | ||
| NM_001300032.1 | NP_001286961.1 | 17 - 18 | 2657 | 63 | ||
| NM_001300033.1 | NP_001286962.1 | 15 - 16 | 2552 | 63 | ||
| NM_176293.3 | NP_788471.1 | 12 - 13 | 2886 | 63 | ||
| NM_176294.5 | NP_788472.2 | 12 - 13 | 2886 | 63 | ||
| GenBank mRNA | AY118310.1 | - | 12 - 13 | 2861 | 63 | |
More Information
Set Membership: |
|

Mp
Genomic Map