Assay Details
| Gene Symbol: Mp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169885.2 | NP_001163356.1 | 19 - 20 | 2911 | 60 | |
| NM_001169886.2 | NP_001163357.1 | 15 - 16 | 3172 | 60 | ||
| NM_001169887.2 | NP_001163358.1 | 21 - 22 | 2961 | 60 | ||
| NM_001169888.2 | NP_001163359.1 | 16 - 17 | 3250 | 60 | ||
| NM_001169889.2 | NP_001163360.1 | 3 - 4 | 1582 | 60 | ||
| NM_001169890.2 | NP_001163361.1 | 14 - 15 | 2108 | 60 | ||
| NM_001169891.2 | NP_001163362.2 | 20 - 21 | 2906 | 60 | ||
| NM_001169892.2 | NP_001163363.1 | 18 - 19 | 2805 | 60 | ||
| NM_001169893.2 | NP_001163364.1 | 20 - 21 | 2943 | 60 | ||
| NM_001259722.1 | NP_001246651.1 | 21 - 22 | 2979 | 60 | ||
| NM_001259723.1 | NP_001246652.1 | 3 - 4 | 368 | 60 | ||
| NM_001300031.1 | NP_001286960.1 | 15 - 16 | 3223 | 60 | ||
| NM_001300032.1 | NP_001286961.1 | 21 - 22 | 2958 | 60 | ||
| NM_001300033.1 | NP_001286962.1 | 17 - 18 | 2811 | 60 | ||
| NM_176293.3 | NP_788471.1 | 14 - 15 | 3145 | 60 | ||
| NM_176294.5 | NP_788472.2 | 16 - 17 | 3187 | 60 | ||
| GenBank mRNA | AY118310.1 | - | 16 - 17 | 3162 | 60 | |
| BT124741.1 | - | 3 - 4 | 361 | 60 | ||
More Information
Set Membership: |
|

Mp
Genomic Map