Assay Details
| Gene Symbol: Mp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169885.2 | NP_001163356.1 | 20 - 21 | 3086 | 61 | |
| NM_001169886.2 | NP_001163357.1 | 16 - 17 | 3347 | 61 | ||
| NM_001169887.2 | NP_001163358.1 | 22 - 23 | 3136 | 61 | ||
| NM_001169888.2 | NP_001163359.1 | 17 - 18 | 3425 | 61 | ||
| NM_001169889.2 | NP_001163360.1 | 4 - 5 | 1757 | 61 | ||
| NM_001169890.2 | NP_001163361.1 | 15 - 16 | 2283 | 61 | ||
| NM_001169891.2 | NP_001163362.2 | 21 - 22 | 3081 | 61 | ||
| NM_001169892.2 | NP_001163363.1 | 19 - 20 | 2980 | 61 | ||
| NM_001169893.2 | NP_001163364.1 | 21 - 22 | 3118 | 61 | ||
| NM_001259722.1 | NP_001246651.1 | 22 - 23 | 3154 | 61 | ||
| NM_001259723.1 | NP_001246652.1 | 4 - 5 | 543 | 61 | ||
| NM_001300031.1 | NP_001286960.1 | 16 - 17 | 3398 | 61 | ||
| NM_001300032.1 | NP_001286961.1 | 22 - 23 | 3133 | 61 | ||
| NM_001300033.1 | NP_001286962.1 | 18 - 19 | 2986 | 61 | ||
| NM_176293.3 | NP_788471.1 | 15 - 16 | 3320 | 61 | ||
| NM_176294.5 | NP_788472.2 | 17 - 18 | 3362 | 61 | ||
| GenBank mRNA | AY118310.1 | - | 17 - 18 | 3337 | 61 | |
| BT124741.1 | - | 4 - 5 | 536 | 61 | ||
More Information
Set Membership: |
|

Mp
Genomic Map