Assay Details
| Gene Symbol: Mp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169885.2 | NP_001163356.1 | 21 - 22 | 3331 | 64 | |
| NM_001169886.2 | NP_001163357.1 | 17 - 18 | 3592 | 64 | ||
| NM_001169887.2 | NP_001163358.1 | 23 - 24 | 3381 | 64 | ||
| NM_001169888.2 | NP_001163359.1 | 18 - 19 | 3670 | 64 | ||
| NM_001169889.2 | NP_001163360.1 | 5 - 6 | 2002 | 64 | ||
| NM_001169890.2 | NP_001163361.1 | 16 - 17 | 2528 | 64 | ||
| NM_001169891.2 | NP_001163362.2 | 22 - 23 | 3326 | 64 | ||
| NM_001169892.2 | NP_001163363.1 | 20 - 21 | 3225 | 64 | ||
| NM_001169893.2 | NP_001163364.1 | 22 - 23 | 3363 | 64 | ||
| NM_001259722.1 | NP_001246651.1 | 23 - 24 | 3399 | 64 | ||
| NM_001259723.1 | NP_001246652.1 | 5 - 6 | 788 | 64 | ||
| NM_001300031.1 | NP_001286960.1 | 17 - 18 | 3643 | 64 | ||
| NM_001300032.1 | NP_001286961.1 | 23 - 24 | 3378 | 64 | ||
| NM_001300033.1 | NP_001286962.1 | 19 - 20 | 3231 | 64 | ||
| NM_176293.3 | NP_788471.1 | 16 - 17 | 3565 | 64 | ||
| NM_176294.5 | NP_788472.2 | 18 - 19 | 3607 | 64 | ||
| GenBank mRNA | AY118310.1 | - | 18 - 19 | 3582 | 64 | |
| BT124741.1 | - | 5 - 6 | 781 | 64 | ||
More Information
Set Membership: |
|

Mp
Genomic Map