Assay Details
| Gene Symbol: Mp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169886.2 | NP_001163357.1 | 2 - 3 | 1451 | 53 | |
| NM_001300031.1 | NP_001286960.1 | 2 - 3 | 1451 | 53 | ||
| NM_176293.3 | NP_788471.1 | 2 - 3 | 1451 | 53 | ||
| NM_176294.5 | NP_788472.2 | 2 - 3 | 1451 | 53 | ||
| GenBank mRNA | AY118310.1 | - | 2 - 3 | 1426 | 53 | |
More Information
Set Membership: |
|

Mp
Genomic Map