Assay Details
| Gene Symbol: Mp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169885.2 | NP_001163356.1 | 8 - 9 | 1285 | 60 | |
| NM_001169886.2 | NP_001163357.1 | 3 - 4 | 1519 | 60 | ||
| NM_001169887.2 | NP_001163358.1 | 8 - 9 | 1290 | 60 | ||
| NM_001169888.2 | NP_001163359.1 | 5 - 6 | 1624 | 60 | ||
| NM_001169891.2 | NP_001163362.2 | 8 - 9 | 1265 | 60 | ||
| NM_001169893.2 | NP_001163364.1 | 8 - 9 | 1290 | 60 | ||
| NM_001259722.1 | NP_001246651.1 | 8 - 9 | 1260 | 60 | ||
| NM_001300031.1 | NP_001286960.1 | 3 - 4 | 1519 | 60 | ||
| NM_001300032.1 | NP_001286961.1 | 8 - 9 | 1290 | 60 | ||
| NM_001300033.1 | NP_001286962.1 | 6 - 7 | 1185 | 60 | ||
| NM_176293.3 | NP_788471.1 | 3 - 4 | 1519 | 60 | ||
| NM_176294.5 | NP_788472.2 | 3 - 4 | 1519 | 60 | ||
| GenBank mRNA | AY118310.1 | - | 3 - 4 | 1494 | 60 | |
More Information
Set Membership: |
|

Mp
Genomic Map