Assay Details
| Gene Symbol: Mp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169885.2 | NP_001163356.1 | 10 - 11 | 1568 | 93 | |
| NM_001169886.2 | NP_001163357.1 | 5 - 6 | 1802 | 93 | ||
| NM_001169887.2 | NP_001163358.1 | 10 - 11 | 1573 | 93 | ||
| NM_001169888.2 | NP_001163359.1 | 7 - 8 | 1907 | 93 | ||
| NM_001169890.2 | NP_001163361.1 | 5 - 6 | 765 | 93 | ||
| NM_001169891.2 | NP_001163362.2 | 10 - 11 | 1548 | 93 | ||
| NM_001169892.2 | NP_001163363.1 | 9 - 10 | 1462 | 93 | ||
| NM_001169893.2 | NP_001163364.1 | 10 - 11 | 1573 | 93 | ||
| NM_001259722.1 | NP_001246651.1 | 10 - 11 | 1543 | 93 | ||
| NM_001300031.1 | NP_001286960.1 | 5 - 6 | 1802 | 93 | ||
| NM_001300032.1 | NP_001286961.1 | 10 - 11 | 1573 | 93 | ||
| NM_001300033.1 | NP_001286962.1 | 8 - 9 | 1468 | 93 | ||
| NM_176293.3 | NP_788471.1 | 5 - 6 | 1802 | 93 | ||
| NM_176294.5 | NP_788472.2 | 5 - 6 | 1802 | 93 | ||
| GenBank mRNA | AY118310.1 | - | 5 - 6 | 1777 | 93 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001169890.2 transcripts.
Set Membership: |
|

Mp
Genomic Map