Assay Details
| Gene Symbol: Mp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169885.2 | NP_001163356.1 | 11 - 12 | 1712 | 57 | |
| NM_001169886.2 | NP_001163357.1 | 6 - 7 | 1946 | 57 | ||
| NM_001169888.2 | NP_001163359.1 | 8 - 9 | 2051 | 57 | ||
| NM_001169890.2 | NP_001163361.1 | 6 - 7 | 909 | 57 | ||
| NM_001169891.2 | NP_001163362.2 | 11 - 12 | 1692 | 57 | ||
| NM_001169892.2 | NP_001163363.1 | 10 - 11 | 1606 | 57 | ||
| NM_001169893.2 | NP_001163364.1 | 11 - 12 | 1717 | 57 | ||
| NM_001259722.1 | NP_001246651.1 | 11 - 12 | 1687 | 57 | ||
| NM_001300031.1 | NP_001286960.1 | 6 - 7 | 1946 | 57 | ||
| NM_001300032.1 | NP_001286961.1 | 11 - 12 | 1717 | 57 | ||
| NM_001300033.1 | NP_001286962.1 | 9 - 10 | 1612 | 57 | ||
| NM_176293.3 | NP_788471.1 | 6 - 7 | 1946 | 57 | ||
| NM_176294.5 | NP_788472.2 | 6 - 7 | 1946 | 57 | ||
| GenBank mRNA | AY118310.1 | - | 6 - 7 | 1921 | 57 | |
More Information
Set Membership: |
|

Mp
Genomic Map