Assay Details
| Gene Symbol: Sxl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031891.3 | NP_001027062.1 | 1 - 2 | 189 | 88 | |
| NM_001031892.2 | NP_001027063.1 | 1 - 2 | 189 | 88 | ||
| NM_001169217.2 | NP_001162688.1 | 1 - 2 | 189 | 88 | ||
| NM_080052.4 | NP_524791.3 | 1 - 2 | 189 | 88 | ||
| GenBank mRNA | BT125904.1 | - | 1 - 2 | 181 | 88 | |
More Information
Set Membership: |
|

Sxl
Genomic Map