Assay Details
| Gene Symbol: Sxl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031893.2 | NP_001027064.1 | 3 - 4 | 692 | 88 | |
| NM_001031895.1 | NP_001027066.1 | 3 - 4 | 778 | 88 | ||
| NM_001169216.2 | NP_001162687.1 | 4 - 5 | 784 | 88 | ||
| NM_001272376.1 | NP_001259305.1 | 3 - 4 | 722 | 88 | ||
| NM_001272377.1 | NP_001259306.1 | 3 - 4 | 740 | 88 | ||
| NM_167113.3 | NP_727163.2 | 3 - 4 | 292 | 88 | ||
| NM_167115.3 | NP_727165.2 | 3 - 4 | 362 | 88 | ||
| NM_167117.2 | NP_727167.2 | 3 - 4 | 692 | 88 | ||
| NM_167118.2 | NP_727168.1 | 3 - 4 | 692 | 88 | ||
| GenBank mRNA | M23635.1 | - | 3 - 4 | 741 | 88 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_167113.3, NM_167115.3 transcripts.
Set Membership: |
|

Sxl
Genomic Map