Assay Details
| Gene Symbol: Sxl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031893.2 | NP_001027064.1 | 2 - 3 | 501 | 103 | |
| NM_001272377.1 | NP_001259306.1 | 2 - 3 | 549 | 103 | ||
| NM_167117.2 | NP_727167.2 | 2 - 3 | 501 | 103 | ||
| NM_167118.2 | NP_727168.1 | 2 - 3 | 501 | 103 | ||
| GenBank mRNA | M23635.1 | - | 2 - 3 | 550 | 103 | |
More Information
Set Membership: |
|

Sxl
Genomic Map