Assay Details
| Gene Symbol: Tbh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298067.1 | NP_001284996.1 | 6 - 7 | 1924 | 65 | |
| NM_176711.3 | NP_788884.1 | 7 - 8 | 1925 | 65 | ||
More Information
Set Membership: |
|

Tbh
Genomic Map