Assay Details
| Gene Symbol: spri | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272484.1 | NP_001259413.1 | 3 - 4 | 1010 | 61 | |
| NM_001272485.1 | NP_001259414.1 | 3 - 4 | 1095 | 61 | ||
| NM_001272489.1 | NP_001259418.1 | 4 - 5 | 2413 | 61 | ||
| NM_176723.3 | NP_788896.1 | 3 - 4 | 1098 | 61 | ||
More Information
Set Membership: |
|

spri
Genomic Map