Assay Details
| Gene Symbol: Vha55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170126.1 | NP_001163597.1 | 3 - 4 | 1453 | 71 | |
| NM_057560.4 | NP_476908.1 | 3 - 4 | 1582 | 71 | ||
| NM_169475.2 | NP_731726.1 | 2 - 3 | 1518 | 71 | ||
| GenBank mRNA | AY051623.1 | - | 3 - 4 | 1453 | 71 | |
| BT001302.1 | - | 2 - 3 | 1518 | 71 | ||
More Information
Set Membership: |
|

Vha55
Genomic Map