Assay Details
| Gene Symbol: Mcm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057773.4 | NP_477121.1 | 1 - 2 | 2265 | 102 | |
| GenBank mRNA | AY069702.1 | - | 1 - 2 | 2191 | 102 | |
| L42762.2 | - | 1 - 2 | 2184 | 102 | ||
More Information
Set Membership: |
|

Mcm2
Genomic Map