Assay Details
| Gene Symbol: Gcn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276212.1 | NP_001263141.1 | 9 - 10 | 4978 | 68 | |
| NM_001276213.1 | NP_001263142.1 | 9 - 10 | 4984 | 68 | ||
| NM_057882.4 | NP_477230.1 | 9 - 10 | 4978 | 68 | ||
| GenBank mRNA | BT126024.1 | - | 9 - 10 | 4954 | 68 | |
More Information
Set Membership: |
|

Gcn2
Genomic Map