Assay Details
| Gene Symbol: Apc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001347814.1 | NP_001334698.1 | 2 - 3 | 781 | 61 | |
| NM_001347815.1 | NP_001334699.1 | 1 - 2 | 841 | 61 | ||
| NM_058082.3 | NP_477430.1 | 1 - 2 | 841 | 61 | ||
| GenBank mRNA | AF091430.2 | - | 1 - 2 | 828 | 61 | |
| AF113913.1 | - | 1 - 2 | 849 | 61 | ||
More Information
Set Membership: |
|

Apc2
Genomic Map