Assay Details
| Gene Symbol: Atu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079516.3 | NP_524240.1 | 2 - 3 | 201 | 68 | |
| GenBank mRNA | AY075403.1 | - | 2 - 3 | 186 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atu| Gene Symbol: Atu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079516.3 | NP_524240.1 | 2 - 3 | 201 | 68 | |
| GenBank mRNA | AY075403.1 | - | 2 - 3 | 186 | 68 | |
Set Membership: |
|