Assay Details
| Gene Symbol: pum | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260073.2 | NP_001247002.1 | 6 - 7 | 2976 | 101 | |
| NM_001275474.1 | NP_001262403.1 | 12 - 13 | 5130 | 101 | ||
| NM_001275475.1 | NP_001262404.1 | 5 - 6 | 2964 | 101 | ||
| NM_079561.4 | NP_524285.2 | 12 - 13 | 5130 | 101 | ||
| NM_169258.3 | NP_731314.1 | 12 - 13 | 4982 | 101 | ||
| NM_169259.3 | NP_731315.1 | 12 - 13 | 4980 | 101 | ||
| NM_169260.4 | NP_731316.2 | 10 - 11 | 3615 | 101 | ||
| NM_176427.3 | NP_788604.1 | 6 - 7 | 3006 | 101 | ||
| GenBank mRNA | BT001872.1 | - | 6 - 7 | 2406 | 101 | |
More Information
Set Membership: |
|

pum
Genomic Map