Assay Details
| Gene Symbol: pum | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275474.1 | NP_001262403.1 | 2 - 3 | 1351 | 95 | |
| NM_079561.4 | NP_524285.2 | 2 - 3 | 1351 | 95 | ||
| NM_169258.3 | NP_731314.1 | 2 - 3 | 1203 | 95 | ||
| NM_169259.3 | NP_731315.1 | 2 - 3 | 1201 | 95 | ||
More Information
Set Membership: |
|

pum
Genomic Map