Assay Details
| Gene Symbol: pum | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275474.1 | NP_001262403.1 | 5 - 6 | 2504 | 98 | |
| NM_079561.4 | NP_524285.2 | 5 - 6 | 2504 | 98 | ||
| NM_169258.3 | NP_731314.1 | 5 - 6 | 2356 | 98 | ||
| NM_169259.3 | NP_731315.1 | 5 - 6 | 2354 | 98 | ||
| NM_169260.4 | NP_731316.2 | 3 - 4 | 989 | 98 | ||
More Information
Set Membership: |
|

pum
Genomic Map