Assay Details
| Gene Symbol: pum | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260073.2 | NP_001247002.1 | 3 - 4 | 1307 | 58 | |
| NM_001275474.1 | NP_001262403.1 | 9 - 10 | 3461 | 58 | ||
| NM_001275475.1 | NP_001262404.1 | 2 - 3 | 1295 | 58 | ||
| NM_079561.4 | NP_524285.2 | 9 - 10 | 3461 | 58 | ||
| NM_169258.3 | NP_731314.1 | 9 - 10 | 3313 | 58 | ||
| NM_169259.3 | NP_731315.1 | 9 - 10 | 3311 | 58 | ||
| NM_169260.4 | NP_731316.2 | 7 - 8 | 1946 | 58 | ||
| NM_176427.3 | NP_788604.1 | 3 - 4 | 1337 | 58 | ||
| GenBank mRNA | BT001872.1 | - | 3 - 4 | 737 | 58 | |
More Information
Set Membership: |
|

pum
Genomic Map