Assay Details
| Gene Symbol: Caf1-55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170146.2 | NP_001163617.1 | 2 - 3 | 899 | 83 | |
| NM_079630.4 | NP_524354.1 | 2 - 3 | 902 | 83 | ||
| GenBank mRNA | AY061408.1 | - | 2 - 3 | 902 | 83 | |
| BT126192.1 | - | 2 - 3 | 626 | 83 | ||
| U62388.1 | - | 2 - 3 | 876 | 83 | ||
More Information
Set Membership: |
|

Caf1-55
Genomic Map