Assay Details
| Gene Symbol: mRpS11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079658.4 | NP_524382.1 | 1 - 2 | 267 | 57 | |
| GenBank mRNA | AY095194.1 | - | 1 - 2 | 267 | 57 | |
| DQ062760.1 | - | 1 - 2 | 201 | 57 | ||
More Information
Set Membership: |
|

mRpS11
Genomic Map