Assay Details
| Gene Symbol: osa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170168.2 | NP_001163639.1 | 3 - 4 | 1481 | 107 | |
| NM_001275751.1 | NP_001262680.1 | 3 - 4 | 1481 | 107 | ||
| NM_001275752.1 | NP_001262681.1 | 3 - 4 | 1772 | 107 | ||
| NM_079668.3 | NP_524392.2 | 3 - 4 | 1772 | 107 | ||
| NM_169775.2 | NP_732263.1 | 3 - 4 | 1772 | 107 | ||
| NM_206506.2 | NP_996228.1 | 3 - 4 | 1772 | 107 | ||
More Information
Set Membership: |
|

osa
Genomic Map