Assay Details
| Gene Symbol: ninaE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079683.3 | NP_524407.1 | 3 - 4 | 897 | 66 | |
| GenBank mRNA | BT010221.1 | - | 3 - 4 | 893 | 66 | |
| BT023714.1 | - | 1841 | 66 | |||
| BT029277.1 | - | 1896 | 66 | |||
More Information
Set Membership: |
|

ninaE
Genomic Map