Assay Details
| Gene Symbol: CCT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079726.4 | NP_524450.2 | 2 - 3 | 333 | 122 | |
| NM_170016.2 | NP_732748.1 | 3 - 4 | 269 | 122 | ||
| GenBank mRNA | AY118416.1 | - | 2 - 3 | 329 | 122 | |
More Information
Set Membership: |
|

CCT1
Genomic Map