Assay Details
| Gene Symbol: gpp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141398.4 | NP_649655.1 | 8 - 9 | 6839 | 63 | |
| NM_169142.2 | NP_731083.1 | 7 - 8 | 7225 | 63 | ||
| NM_169143.2 | NP_731084.1 | 8 - 9 | 6839 | 63 | ||
More Information
Set Membership: |
|

gpp
Genomic Map