Assay Details
| Gene Symbol: MAGE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141445.3 | NP_649702.2 | 1 | 805 | 139 | |
| GenBank mRNA | AY089612.1 | - | 1 | 801 | 139 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MAGE| Gene Symbol: MAGE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141445.3 | NP_649702.2 | 1 | 805 | 139 | |
| GenBank mRNA | AY089612.1 | - | 1 | 801 | 139 | |
Set Membership: |
|