Assay Details
| Gene Symbol: f-cup | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275615.1 | NP_001262544.1 | 3 - 4 | 569 | 69 | |
| NM_001275616.1 | NP_001262545.1 | 1 - 2 | 481 | 69 | ||
| NM_001275617.1 | NP_001262546.1 | 1 - 2 | 481 | 69 | ||
| NM_142049.3 | NP_650306.1 | 1 - 2 | 481 | 69 | ||
| NM_169550.2 | NP_731852.2 | 3 - 4 | 569 | 69 | ||
| GenBank mRNA | AF184226.1 | - | 1 - 2 | 453 | 69 | |
More Information
Set Membership: |
|

f-cup
Genomic Map