Assay Details
| Gene Symbol: f-cup | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104302.2 | NP_001097772.1 | 5 - 6 | 1272 | 89 | |
| NM_001104303.2 | NP_001097773.1 | 3 - 4 | 399 | 89 | ||
| NM_001104304.2 | NP_001097774.1 | 2 - 3 | 416 | 89 | ||
| NM_001275615.1 | NP_001262544.1 | 4 - 5 | 755 | 89 | ||
| NM_001275616.1 | NP_001262545.1 | 2 - 3 | 667 | 89 | ||
| NM_001275617.1 | NP_001262546.1 | 2 - 3 | 667 | 89 | ||
| NM_001275618.1 | NP_001262547.1 | 3 - 4 | 1184 | 89 | ||
| NM_142049.3 | NP_650306.1 | 2 - 3 | 667 | 89 | ||
| NM_169550.2 | NP_731852.2 | 4 - 5 | 755 | 89 | ||
| GenBank mRNA | AF184226.1 | - | 2 - 3 | 639 | 89 | |
| BT012334.1 | - | 5 - 6 | 1221 | 89 | ||
More Information
Set Membership: |
|

f-cup
Genomic Map