Assay Details
| Gene Symbol: f-cup | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104304.2 | NP_001097774.1 | 6 - 7 | 1323 | 76 | |
| NM_142049.3 | NP_650306.1 | 6 - 7 | 1574 | 76 | ||
| NM_169550.2 | NP_731852.2 | 8 - 9 | 1662 | 76 | ||
| GenBank mRNA | AF184226.1 | - | 6 - 7 | 1546 | 76 | |
More Information
Set Membership: |
|

f-cup
Genomic Map