Assay Details
| Gene Symbol: f-cup | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104302.2 | NP_001097772.1 | 11 - 12 | 2654 | 90 | |
| NM_001104303.2 | NP_001097773.1 | 9 - 10 | 1779 | 90 | ||
| NM_001104304.2 | NP_001097774.1 | 7 - 8 | 1466 | 90 | ||
| NM_001275615.1 | NP_001262544.1 | 10 - 11 | 2132 | 90 | ||
| NM_001275616.1 | NP_001262545.1 | 8 - 9 | 2044 | 90 | ||
| NM_001275617.1 | NP_001262546.1 | 8 - 9 | 2042 | 90 | ||
| NM_001275618.1 | NP_001262547.1 | 9 - 10 | 2566 | 90 | ||
| NM_142049.3 | NP_650306.1 | 7 - 8 | 1717 | 90 | ||
| NM_169550.2 | NP_731852.2 | 9 - 10 | 1805 | 90 | ||
| GenBank mRNA | AF184226.1 | - | 7 - 8 | 1689 | 90 | |
| BT012334.1 | - | 11 - 12 | 2603 | 90 | ||
More Information
Set Membership: |
|

f-cup
Genomic Map