Assay Details
| Gene Symbol: f-cup | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104302.2 | NP_001097772.1 | 12 - 13 | 2837 | 74 | |
| NM_001104303.2 | NP_001097773.1 | 10 - 11 | 1962 | 74 | ||
| NM_001104304.2 | NP_001097774.1 | 8 - 9 | 1649 | 74 | ||
| NM_001275615.1 | NP_001262544.1 | 11 - 12 | 2315 | 74 | ||
| NM_001275616.1 | NP_001262545.1 | 9 - 10 | 2227 | 74 | ||
| NM_001275617.1 | NP_001262546.1 | 9 - 10 | 2225 | 74 | ||
| NM_001275618.1 | NP_001262547.1 | 10 - 11 | 2749 | 74 | ||
| NM_142049.3 | NP_650306.1 | 8 - 9 | 1900 | 74 | ||
| NM_169550.2 | NP_731852.2 | 10 - 11 | 1988 | 74 | ||
| GenBank mRNA | AF184226.1 | - | 8 - 9 | 1872 | 74 | |
| BT012334.1 | - | 12 - 13 | 2786 | 74 | ||
More Information
Set Membership: |
|

f-cup
Genomic Map