Assay Details
| Gene Symbol: natalisin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104319.2 | NP_001097789.1 | 1 | 470 | 58 | |
| NM_001170137.1 | NP_001163608.1 | 1 | 470 | 58 | ||
| GenBank mRNA | BT023179.1 | - | 1 | 456 | 58 | |
More Information
Set Membership: |
|

natalisin
Genomic Map