Assay Details
| Gene Symbol: UQCR-C1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142144.3 | NP_650401.1 | 6 - 7 | 1121 | 100 | |
| NM_169607.2 | NP_731954.1 | 5 - 6 | 1193 | 100 | ||
| GenBank mRNA | AY058243.1 | - | 5 - 6 | 1074 | 100 | |
More Information
Set Membership: |
|

UQCR-C1
Genomic Map